Tantalum pentafluoride 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC6630-25G
PC6630-5G
APOSPC6630-25GEA
17390
CZK
APOSPC6630-25G
APOSPC6630-5G
Tantalum pentafluoride 99%
Tantalum pentafluoride
Vzorec:
F₅Ta Bod varu: 229,5 °C Bod tání: 97 °C Hustota: 4,74 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042542 CAS číslo: 7783-71-3 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...