Tisk…

2-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-5-(trifluoromethyl)benzene-1-sulphonyl chloride

PC6582-25G PC6582-5G
APOSPC6582-25GEA 4140 CZK
APOSPC6582-25G APOSPC6582-5G
2-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride
2-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride
Vzorec: C₇H₃BrClF₃O₂S
MW: 323,52 g/mol
Bod varu: 230…231 °C
Hustota: 1,854 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03094395
CAS číslo: 176225-08-4
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR