2-Bromo-4-(trifluoromethyl)benzenesulphonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-4-(trifluoromethyl)benzene-1-sulphonyl chloride
PC6581-25G
PC6581-5G
APOSPC6581-25GEA
4960
CZK
APOSPC6581-25G
APOSPC6581-5G
2-Bromo-4-(trifluoromethyl)benzenesulphonyl chloride 97%
2-Bromo-4-(trifluoromethyl)benzenesulphonyl chloride
Vzorec:
C₇H₃BrClF₃O₂S MW: 323,52 g/mol Bod varu: 235…236 °C Hustota: 1,865 g/cm³ (25 °C) Bod vzplanutí: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094394 CAS číslo: 54403-98-4 UN: 2920 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...