Tisk…

Tin(IV) fluoride 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC6580-1G PC6580-5G
APOSPC6580-1GEA 3230 CZK
APOSPC6580-1G APOSPC6580-5G
Tin(IV) fluoride 98%
Tin(IV) fluoride
Vzorec: SnF₄
MW: 194.7
Bod varu: 705
Bod tání: 705 °C (subl.)
Hustota: 4.78
Storage Temperature: Ambient
MDL Number: MFCD00042541
CAS číslo: 7783-62-2
EINECS: 232-016-0
UN: 2923
ADR: 8+6.1 III
Merck Index: 12,08931

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR