4-Bromo-3-(trifluoromethyl)benzenesulphonyl chloride 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
4-Bromo-3-(trifluoromethyl)benzene-1-sulphonyl chloride
PC6575-100G
PC6575-25G
PC6575-5G
APOSPC6575-100GEA
16170
CZK
APOSPC6575-100G
APOSPC6575-25G
APOSPC6575-5G
4-Bromo-3-(trifluoromethyl)benzenesulphonyl chloride 98%
4-Bromo-3-(trifluoromethyl)benzenesulphonyl chloride
Vzorec:
C₇H₃BrClF₃O₂S MW: 323,52 g/mol Hustota: 1,833 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094391 CAS číslo: 351003-47-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...