Tisk…

3-(2-Fluorophenoxy)benzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3'-Bromomethyl-2-fluorodiphenyl Ether

PC6515-100MG PC6515-250MG
APOSPC6515-100MGEA 3640 CZK
APOSPC6515-100MG APOSPC6515-250MG
3-(2-Fluorophenoxy)benzyl bromide
3-(2-Fluorophenoxy)benzyl bromide
Vzorec: C₁₃H₁₀BrFO
MW: 281,12 g/mol
Hustota: 1,46 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00059914
CAS číslo: 242812-04-0
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR