Tisk…

Sodium hexafluorotitanate 98%

Dodavatel: Apollo Scientific
Varování

PC6514-5G PC6514-10G PC6514-50G
APOSPC6514-5GEA 2070 CZK
APOSPC6514-5G APOSPC6514-10G APOSPC6514-50G
Sodium hexafluorotitanate 98%
Sodium hexafluorotitanate
Vzorec: F₆Na₂Ti
Storage Temperature: Ambient
MDL Number: MFCD00014244
CAS číslo: 17116-13-1
EINECS: 241-181-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR