Sodium heptafluorobutyrate 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC6470-5G
PC6470-25G
APOSPC6470-5GEA
1950
CZK
APOSPC6470-5G
APOSPC6470-25G
Sodium heptafluorobutyrate 98%
Sodium heptafluorobutyrate
Vzorec:
C₄F₇NaO₂ MW: 236,02 g/mol Bod tání: 248…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066168 CAS číslo: 2218-54-4 EINECS: 218-721-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...