Silver trifluoroacetate 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC6440-25G
PC6440-5G
PC6440-100G
APOSPC6440-25GEA
1490
CZK
APOSPC6440-25G
APOSPC6440-5G
APOSPC6440-100G
Silver trifluoroacetate 98%
Silver trifluoroacetate
Lewis acid catalyst for regioselective alkylation:Helv.Chim.Acta., 1989, 72, 1362; Can convert enol silyl ethers to alpha-acyloxy carbonyls, ask for reference.
Vzorec:
C₂AgF₃O₂ MW: 220,88 g/mol Bod tání: 257…260 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00013199 CAS číslo: 2966-50-9 EINECS: 221-004-0 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...