Tisk…

4-Methoxy-2-(trifluoromethoxy)benzyl bromide 97%

Dodavatel: Apollo Scientific

PC6389-10G PC6389-5G PC6389-1G
APOSPC6389-10GEA 11540 CZK
APOSPC6389-10G APOSPC6389-5G APOSPC6389-1G
4-Methoxy-2-(trifluoromethoxy)benzyl bromide 97%
4-Methoxy-2-(trifluoromethoxy)benzyl bromide
Vzorec: C₉H₈BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD06660289
CAS číslo: 886502-56-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR