Tisk…

Methyl-2-fluoro-5-iodobenzoate

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl 2-Fluoro-5-iodobenzoate

PC6375-1G PC6375-5G PC6375-25G
APOSPC6375-1GEA 1040 CZK
APOSPC6375-1G APOSPC6375-5G APOSPC6375-25G
Methyl-2-fluoro-5-iodobenzoate
Methyl-2-fluoro-5-iodobenzoate
Vzorec: C₈H₆FIO₂
MW: 280,0349 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203930
CAS číslo: 625471-27-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR