Tisk…

Silver(II) fluoride 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Silver difluoride, Silver(II)fluoride

PC6360-10G PC6360-5G
APOSPC6360-10GEA 9660 CZK
APOSPC6360-10G APOSPC6360-5G
Silver(II) fluoride 99%
Silver(II) fluoride

Hard fluorinating agent; Liberates iodine from KI solution. Can be used for substitution of hydrogens in hydrohalogenate alkenes

Vzorec: AgF₂
Bod tání: 690 °C
Hustota: 4,57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003411
CAS číslo: 7783-95-1
EINECS: 232-037-5
UN: 3085
ADR: 5.1+8,II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR