8-Fluoro-4-quinolinol ≥95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
8-Fluoroquinolin-4-ol, 8-Fluoro-4-hydroxyquinoline
PC6349-1G
PC6349-250MG
APOSPC6349-1GEA
622
CZK
APOSPC6349-1G
APOSPC6349-250MG
8-Fluoro-4-quinolinol ≥95%
8-Fluoro-4-quinolinol
Vzorec:
C₉H₆FNO MW: 163.15 g/mol Bod tání: 278…282 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272395 CAS číslo: 63010-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...