Tisk…

Scandium(III) fluoride 99,9%

Dodavatel: Apollo Scientific
Nebezpečí

PC6343R-10G PC6343R-5G
APOSPC6343R-10GEA 12960 CZK
APOSPC6343R-10G APOSPC6343R-5G
Scandium(III) fluoride 99,9%
Scandium(III) fluoride
Vzorec: F₃Sc
MW: 101.95
Bod varu: 1527
Bod tání: 1515 °C
Hustota: 2.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016325
CAS číslo: 13709-47-2
EINECS: 237-255-4
UN: 3288
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR