Samarium(III) fluoride 99,9%
Dodavatel: Apollo Scientific
PC6343-100G
PC6343-25G
APOSPC6343-100GEA
21690
CZK
APOSPC6343-100G
APOSPC6343-25G
Samarium(III) fluoride 99,9%
Samarium(III) fluoride
Vzorec:
F₃Sm MW: 207.36 Bod varu: 2323 °C Bod tání: 1306 °C Hustota: 6.928 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00049540 CAS číslo: 13765-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...