Tisk…

2-(4-Chlorophenyl)-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile 97%

Dodavatel: Apollo Scientific

PC6335-1G PC6335-500MG PC6335-5G
APOSPC6335-1GEA 7840 CZK
APOSPC6335-1G APOSPC6335-500MG APOSPC6335-5G
2-(4-Chlorophenyl)-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile 97%
2-(4-Chlorophenyl)-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]acetonitrile
Vzorec: C₁₄H₇Cl₂F₃N₂
MW: 331,13 g/mol
Bod varu: 146 °C/0,1mmHg
Bod tání: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00172211
CAS číslo: 213993-80-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR