Tisk…

Potassium trifluoromethanesulphonate 99%

Dodavatel: Apollo Scientific
Varování

PC6324-25G PC6324-5G
APOSPC6324-25GEA 6650 CZK
APOSPC6324-25G APOSPC6324-5G
Potassium trifluoromethanesulphonate 99%
Potassium trifluoromethanesulphonate
Vzorec: CF₃KO₃S
Bod tání: 236 °C
Storage Temperature: Ambient
MDL Number: MFCD00042370
CAS číslo: 2926-27-4
EINECS: 608-334-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR