Tisk…

5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Chloro-1,6-dihydro-6-oxo-1-[4-(trifluoromethyl)benzyl]pyridine-3-carboxylic acid

PC6322-1G PC6322-250MG
APOSPC6322-1GEA 14780 CZK
APOSPC6322-1G APOSPC6322-250MG
5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid 97%
5-Chloro-6-oxo-1-[4-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid
Vzorec: C₁₄H₉ClF₃NO₃
MW: 331,68 g/mol
Bod tání: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD04038241
CAS číslo: 886761-94-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR