5-Chloro-4-cyano-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonitrile
PC6317-1G
PC6317-500MG
APOSPC6317-1GEA
14520
CZK
APOSPC6317-1G
APOSPC6317-500MG
5-Chloro-4-cyano-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%
5-Chloro-4-cyano-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Vzorec:
C₆H₃ClF₃N₃ MW: 209,56 g/mol Bod tání: 84…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD00140981 CAS číslo: 219568-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...