4-(Chloromethyl)-5-(2,4-dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%
Dodavatel: Apollo Scientific
PC6315-1G
PC6315-500MG
APOSPC6315-1GEA
18980
CZK
APOSPC6315-1G
APOSPC6315-500MG
4-(Chloromethyl)-5-(2,4-dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%
4-(Chloromethyl)-5-(2,4-dichlorophenoxy)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Vzorec:
C₁₂H₈Cl₃F₃N₂O Bod tání: 54…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00664860 CAS číslo: 318959-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...