Tisk…

Potassium hexafluorophosphate 99%

Dodavatel: Apollo Scientific
Nebezpečí

PC6290-1KG PC6290-500G PC6290-250G PC6290-100G
APOSPC6290-1KGEA 12070 CZK
APOSPC6290-1KG APOSPC6290-500G APOSPC6290-250G APOSPC6290-100G
Potassium hexafluorophosphate 99%
Potassium hexafluorophosphate

Soluble neutral salt that can be used for synthesis of other hexafluorophosphate salts as a substitutefor hexafluorophosphoric acid

Vzorec: F₆KP
MW: 184.06
Bod tání: 575 °C
Storage Temperature: Ambient
MDL Number: MFCD00011412
CAS číslo: 17084-13-8
EINECS: 241-143-0
UN: 3260
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR