N-(Boc)-7-fluoroindole-2-boronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
7-Fluoro-1H-indole-2-boronic acid, N-BOC protected
PC6246-1G
PC6246-250MG
PC6246-500MG
APOSPC6246-1GEA
35050
CZK
APOSPC6246-1G
APOSPC6246-250MG
APOSPC6246-500MG
N-(Boc)-7-fluoroindole-2-boronic acid
N-(Boc)-7-fluoroindole-2-boronic acid
Vzorec:
C₁₃H₁₅BFNO₄ MW: 279.08 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11109422 CAS číslo: 1000068-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...