5-(3-(Trifluoromethyl)phenyl)oxazole
Dodavatel: Apollo Scientific
|
Varování
|
PC6225-1G
PC6225-5G
APOSPC6225-1GEA
1040
CZK
APOSPC6225-1G
APOSPC6225-5G
5-(3-(Trifluoromethyl)phenyl)oxazole
5-(3-(Trifluoromethyl)phenyl)oxazole
Vzorec:
C₁₀H₆F₃NO MW: 213,1600 g/mol Bod varu: 78 °C/0,6mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD00085180 CAS číslo: 175205-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...