Tisk…

Tris(perfluorobutyl)amine

Dodavatel: Apollo Scientific
Varování

Synonyms: Perfluorotri-n-butylamine, Heptacosafluorotri-n-butylamine, Perfluorotributylamine, FC-43, Heptacosafluorotributylamine, PFTBA, Tris(nonafluorobutyl)amine

PC6220-100G PC6220-25G
APOSPC6220-100GEA 2490 CZK
APOSPC6220-100G APOSPC6220-25G
Tris(perfluorobutyl)amine
Tris(perfluorobutyl)amine
Vzorec: C₁₂F₂₇N
MW: 671,1 g/mol
Bod varu: 170…180 °C
Hustota: 1,9 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000436
CAS číslo: 311-89-7
EINECS: 206-223-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR