Tisk…

Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetra-oxapentadecanoic acid, methyl ester 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: Methyl perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetraoxapentadecanoate

PC6219D-25G PC6219D-5G PC6219D-1G
APOSPC6219D-25GEA 15710 CZK
APOSPC6219D-25G APOSPC6219D-5G APOSPC6219D-1G
Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetra-oxapentadecanoic acid, methyl ester 97%
Perfluoro-2,5,8,11-tetramethyl-3,6,9,12-tetra-oxapentadecanoic acid, methyl ester
Vzorec: C₁₆H₃F₂₉O₆
MW: 842,15 g/mol
Bod varu: 70…80 °C/0,1mmHg
Hustota: 1,74 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00054670
CAS číslo: 133609-46-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR