Tisk…

Hexadecafluorosebacic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Perfluorosebacic Acid, Perfluorodecanedioic Acid, Hexadecafluorodecanedioic Acid

PC6212-25G PC6212-5G
APOSPC6212-25GEA 4960 CZK
APOSPC6212-25G APOSPC6212-5G
Hexadecafluorosebacic acid 95%
Hexadecafluorosebacic acid
Vzorec: C₁₀H₂F₁₆O₄
MW: 490.10 g/mol
Bod varu: 225 °C/60mmHg
Bod tání: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00042365
CAS číslo: 307-78-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR