3-Fluoro-5-hydroxybenzoic acid 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC6191-1G
PC6191-5G
PC6191-10G
APOSPC6191-1GEA
622
CZK
APOSPC6191-1G
APOSPC6191-5G
APOSPC6191-10G
3-Fluoro-5-hydroxybenzoic acid 98%
3-Fluoro-5-hydroxybenzoic acid
Vzorec:
C₇H₅FO₃ MW: 156.1112 g/mol Bod tání: 214…215 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11846515 CAS číslo: 860296-12-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...