Tisk…

3-Amino-2-fluorobenzoic acid 98%

Dodavatel: Apollo Scientific
Varování

PC6187-10G PC6187-1G PC6187-5G
APOSPC6187-10GEA 0 CZK
APOSPC6187-10G APOSPC6187-1G APOSPC6187-5G
3-Amino-2-fluorobenzoic acid 98%
3-Amino-2-fluorobenzoic acid
Vzorec: C7H6FNO2
MW: 155,1300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06797827
CAS číslo: 914223-43-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR