1H,1H,2H,2H-Perfluorooctyltrimethoxysilane 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC6181E-25G
PC6181E-5G
APOSPC6181E-25GEA
9760
CZK
APOSPC6181E-25G
APOSPC6181E-5G
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane 97%
1H,1H,2H,2H-Perfluorooctyltrimethoxysilane
Vzorec:
C₁₁H₁₃F₁₃O₃Si MW: 468,2800 g/mol Bod varu: 112 °C/16mmHg Hustota: 1,457 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00153261 CAS číslo: 85857-16-5 EINECS: 288-657-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...