3-Cyano-4-fluorobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC6131-1G
PC6131-5G
PC6131-25G
APOSPC6131-1GEA
870
CZK
APOSPC6131-1G
APOSPC6131-5G
APOSPC6131-25G
3-Cyano-4-fluorobenzoic acid
3-Cyano-4-fluorobenzoic acid
Vzorec:
C8H4FNO2 MW: 165,12 g/mol Bod tání: 187…188 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094339 CAS číslo: 171050-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...