Tisk…

Nonafluoro(trifluoromethyl)cyclopentane 90%

Dodavatel: Apollo Scientific
Varování

PC6113-100G PC6113-25G PC6113-5G
APOSPC6113-100GEA 4600 CZK
APOSPC6113-100G APOSPC6113-25G APOSPC6113-5G
Nonafluoro(trifluoromethyl)cyclopentane 90%
Nonafluoro(trifluoromethyl)cyclopentane
Vzorec: C₆F₁₂
Bod varu: 47…53 °C
Bod tání: –90 °C
Hustota: 1,68 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066611
CAS číslo: 1805-22-7
EINECS: 217-298-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR