Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate
Dodavatel: Apollo Scientific
Synonyms:
Methyl 5-iodo-3-methyl-2-[(trifluoroacetyl)amino]benzoate
PC6102-1G
PC6102-500MG
PC6102-5G
APOSPC6102-1GEA
4570
CZK
APOSPC6102-1G
APOSPC6102-500MG
APOSPC6102-5G
Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate
Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate
Vzorec:
C₁₁H₉F₃INO₃ MW: 387,1 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD12827837 CAS číslo: 1186405-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...