Tisk…

1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate) 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC6095-25G PC6095-5G
APOSPC6095-25GEA 18720 CZK
APOSPC6095-25G APOSPC6095-5G
1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate) 98%
1,1'-Difluoro-2,2'-bipyridinium bis(tetrafluoroborate)

Fluorinating agent (103g/Kg effective F2), ask for literature

Vzorec: C₁₀H₈B₂F₁₀N₂
MW: 367,79 g/mol
Bod tání: 166…175 °C (rozkl.)
Storage Temperature: Refrigerator
MDL Number: MFCD01073546
CAS číslo: 178439-26-4
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR