3-Bromo-2,5-difluoropyridine 98%
Dodavatel: Apollo Scientific
PC6076-1G
PC6076-250MG
PC6076-5G
APOSPC6076-1GEA
7540
CZK
APOSPC6076-1G
APOSPC6076-250MG
APOSPC6076-5G
3-Bromo-2,5-difluoropyridine 98%
3-Bromo-2,5-difluoropyridine
Vzorec:
C₅H₂BrF₂N Storage Temperature: Ambient |
MDL Number:
MFCD12827553 CAS číslo: 1211331-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...