Tisk…

N-(2-Acetyl-4-iodo-6-methylphenyl)-2,2,2-trifluoroacetamide

Dodavatel: Apollo Scientific

PC6056-1G PC6056-500MG PC6056-5G
APOSPC6056-1GEA 6220 CZK
APOSPC6056-1G APOSPC6056-500MG APOSPC6056-5G
N-(2-Acetyl-4-iodo-6-methylphenyl)-2,2,2-trifluoroacetamide
N-(2-Acetyl-4-iodo-6-methylphenyl)-2,2,2-trifluoroacetamide
Vzorec: C₁₁H₉F₃INO₂
Storage Temperature: Ambient
MDL Number: MFCD12827838
CAS číslo: 935292-71-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR