Tisk…

Perfluoroheptanoic acid 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Tridecafluoroenanthic acid, Tridecafluoroheptylic acid, Tridecafluoroheptanoic acid, Perfluoroenanthic acid

PC6034-100G PC6034-25G
APOSPC6034-100GEA 4500 CZK
APOSPC6034-100G APOSPC6034-25G
Perfluoroheptanoic acid 98%
Perfluoroheptanoic acid
Vzorec: C₇HF₁₃O₂
Bod varu: 175 °C
Bod tání: 30 °C
Hustota: 1,792 g/cm³ (25 °C)
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00039604
CAS číslo: 375-85-9
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR