3,5-Difluoro-4-methoxybenzyl bromide 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC6033-5G
PC6033-1G
PC6033-25G
APOSPC6033-5GEA
6550
CZK
APOSPC6033-5G
APOSPC6033-1G
APOSPC6033-25G
3,5-Difluoro-4-methoxybenzyl bromide 98%
3,5-Difluoro-4-methoxybenzyl bromide
Vzorec:
C₈H₇BrF₂O MW: 237.04 g/mol Bod tání: 39…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115916 CAS číslo: 706786-42-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...