Perfluoroheptane (mixture of isomers) 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC6010-100G
PC6010-25G
PC6010-5G
APOSPC6010-100GEA
3710
CZK
APOSPC6010-100G
APOSPC6010-25G
APOSPC6010-5G
Perfluoroheptane (mixture of isomers) 97%
Perfluoroheptane (mixture of isomers)
Vzorec:
C₇F₁₆ Bod varu: 80…84 °C Hustota: 1,745 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00040339 CAS číslo: 335-57-9 EINECS: 206-392-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...