Tisk…

2,3,5-Trifluorobenzonitrile 98%

Dodavatel: Apollo Scientific
Nebezpečí

PC6009-1G PC6009-25G PC6009-5G
APOSPC6009-1GEA 622 CZK
APOSPC6009-1G APOSPC6009-25G APOSPC6009-5G
2,3,5-Trifluorobenzonitrile 98%
2,3,5-Trifluorobenzonitrile
Vzorec: C₇H₂F₃N
MW: 157,09 g/mol
Hustota: 1,36 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00083539
CAS číslo: 241154-09-6
UN: 3276
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR