Tisk…

(Perfluoro-n-decyl)ethane 97%

Dodavatel: Apollo Scientific
Varování

PC5969M-100MG PC5969M-1G PC5969M-250MG
APOSPC5969M-100MGEA 2940 CZK
APOSPC5969M-100MG APOSPC5969M-1G APOSPC5969M-250MG
(Perfluoro-n-decyl)ethane 97%
(Perfluoro-n-decyl)ethane
Vzorec: C₁₂H₅F₂₁
Bod varu: 90 °C/20mmHg
Hustota: 1,703 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236699
CAS číslo: 154478-87-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR