Tisk…

2,5-Bis(trifluoromethyl)benzyl alcohol 98%

Dodavatel: Apollo Scientific

PC5967-25G PC5967-5G
APOSPC5967-25GEA 8830 CZK
APOSPC5967-25G APOSPC5967-5G
2,5-Bis(trifluoromethyl)benzyl alcohol 98%
2,5-Bis(trifluoromethyl)benzyl alcohol
Vzorec: C₉H₆F₆O
MW: 244,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00799484
CAS číslo: 302911-97-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR