Tisk…

trans-Perfluorodecahydronaphthalene 97%

Dodavatel: Apollo Scientific

Synonyms: trans-Perfluorodecalin

PC5960B-10G PC5960B-25G
APOSPC5960B-10GEA 4370 CZK
APOSPC5960B-10G APOSPC5960B-25G
trans-Perfluorodecahydronaphthalene 97%
trans-Perfluorodecahydronaphthalene
Vzorec: C₁₀F₁₈
Storage Temperature: Ambient
MDL Number: MFCD00153232
CAS číslo: 60433-12-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR