2-Fluoro-4-methylbenzenesulphonyl chloride
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Fluoro-4-methylbenzene-1-sulphonyl chloride
PC5941-25G
PC5941-5G
PC5941-100G
APOSPC5941-25GEA
6580
CZK
APOSPC5941-25G
APOSPC5941-5G
APOSPC5941-100G
2-Fluoro-4-methylbenzenesulphonyl chloride
2-Fluoro-4-methylbenzenesulphonyl chloride
Vzorec:
C₇H₆ClFO₂S MW: 208,64 g/mol Bod tání: 42…44 °C Bod vzplanutí: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094330 CAS číslo: 518070-29-6 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...