5-Fluoro-m-toluidine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Fluoro-5-methylaniline
PC5923-1G
PC5923-5G
PC5923-25G
APOSPC5923-1GEA
622
CZK
APOSPC5923-1G
APOSPC5923-5G
APOSPC5923-25G
5-Fluoro-m-toluidine 97%
5-Fluoro-m-toluidine
Vzorec:
C₇H₈FN MW: 125.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077538 CAS číslo: 52215-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...