Tisk…

(3-Fluoro-5-methylphenyl)acetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(3-Fluoro-5-methylphenyl)acetic acid

PC5921-1G PC5921-5G PC5921-10G
APOSPC5921-1GEA 1490 CZK
APOSPC5921-1G APOSPC5921-5G APOSPC5921-10G
(3-Fluoro-5-methylphenyl)acetic acid
(3-Fluoro-5-methylphenyl)acetic acid
Vzorec: C₉H₉FO₂
MW: 168,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094318
CAS číslo: 518070-22-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR