5-Fluoro-m-toluic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC5915-100G
PC5915-25G
PC5915-5G
APOSPC5915-100GEA
5430
CZK
APOSPC5915-100G
APOSPC5915-25G
APOSPC5915-5G
5-Fluoro-m-toluic acid 98%
5-Fluoro-m-toluic acid
Vzorec:
C₈H₇FO₂ MW: 154.14 Bod tání: 141…145 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094313 CAS číslo: 518070-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...