2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Perfluorobutyl-1,2-epoxypropane
PC5906P-1G
PC5906P-25G
PC5906P-100G
APOSPC5906P-1GEA
622
CZK
APOSPC5906P-1G
APOSPC5906P-25G
APOSPC5906P-100G
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane
Vzorec:
C₇H₅F₉O MW: 276,10 g/mol Bod varu: 52 °C/41mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00236085 CAS číslo: 81190-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...