Perfluoro-2-butene 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
FC-1318my
PC5904-100G
PC5904-25G
PC5904-5G
APOSPC5904-100GEA
39020
CZK
APOSPC5904-100G
APOSPC5904-25G
APOSPC5904-5G
Perfluoro-2-butene 97%
Perfluoro-2-butene
Mixture of cis/trans isomers; major impurity is cis/trans-2H-heptafluoro-2-butene.
Vzorec:
C₄F₈ MW: 200.03 g/mol Bod varu: 1.2 °C Bod tání: –136…–134 °C Hustota: 1.5297 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS číslo:
360-89-4 EINECS: 206-640-9 UN: 2422 ADR: 2.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...