Tisk…

Potassium(pyrrolidin-1-ylmethyl)trifluoroborate

Dodavatel: Apollo Scientific
Varování

Synonyms: Potassium trifluoro[(pyrrolidin-1-yl)methyl]borate

PC5891-1G PC5891-5G PC5891-25G
APOSPC5891-1GEA 3410 CZK
APOSPC5891-1G APOSPC5891-5G APOSPC5891-25G
Potassium(pyrrolidin-1-ylmethyl)trifluoroborate
Potassium(pyrrolidin-1-ylmethyl)trifluoroborate
Vzorec: C₅H₁₀BF₃KN
MW: 191,0441 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10700161
CAS číslo: 888711-53-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR