Perfluorobutane-1-sulphonyl fluoride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride
PC5887-100G
PC5887-250G
PC5887-25G
APOSPC5887-100GEA
1490
CZK
APOSPC5887-100G
APOSPC5887-250G
APOSPC5887-25G
Perfluorobutane-1-sulphonyl fluoride 95%
Perfluorobutane-1-sulphonyl fluoride
Vzorec:
C₄F₁₀O₂S MW: 302.09 g/mol Bod varu: 64…66 °C Bod tání: –110 °C Hustota: 1.682 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007422 CAS číslo: 375-72-4 EINECS: 206-792-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...